Få mere at vide
Chitotetraose Tetradecaacetate, TRC
Artikelnummer. 30368433
Skift visning
Mængde:
2 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368433 5 mg
30367995 10 mg
30351706 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30368433 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3150055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 117399-51-6
Molekylær formel C52 H74 N4 O31
Molekylvægt (g/mol) 1251.15
IUPAC navn [(2R,3S,4R,5R,6S)-5-acetamido-6-[(2R,3S,4R,5R,6S)-5-acetamido-6-[(2R,3S,4R,5R,6S)-5-acetamido-4,6-diacetyloxy-2-(acetyloxymethyl)oxan-3-yl]oxy-4-acetyloxy-2-(acetyloxymethyl)oxan-3-yl]oxy-3-[(2S,3R,4R,5S,6R)-3-acetamido-4,5-diacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxy-4-acetyloxyoxan-2-yl]methyl acetate
SMIL CC(=O)N[C@H]1[C@H](O[C@@H]2[C@@H](COC(=O)C)O[C@@H](O[C@@H]3[C@@H](COC(=O)C)O[C@@H](OC(=O)C)[C@H](NC(=O)C)[C@H]3OC(=O)C)[C@H](NC(=O)C)[C@H]2OC(=O)C)O[C@H](COC(=O)C)[C@@H](O[C@@H]4O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]4NC(=O)C)[C@@H]1OC(=O)C
Tekniske data
Kemisk navn eller materiale Chitotetraose Tetradecaacetate
Mængde 5 mg
InChI formel InChI=1S/C52H74N4O31/c1-19(57)53-37-46(77-29(11)67)42(34(16-72-24(6)62)81-49(37)80-32(14)70)85-51-39(55-21(3)59)48(79-31(13)69)44(36(83-51)18-74-26(8)64)87-52-40(56-22(4)60)47(78-30(12)68)43(35(84-52)17-73-25(7)63)86-50-38(54-20(2)58)45(76-28(10)66)41(75-27(9)65)33(82-50)15-71-23(5)61/h33-52H,15-18H2,1-14H3,(H,53,57)(H,54,58)(H,55,59)(H,56,60)/t33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-,49-,50+,51+,52+/m1/s1
Formel vægt 1250.4337