Få mere at vide
Chitobiose Dihydrochloride, TRC
Artikelnummer. 30136644
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136644 2 mg
30134664 1 mg
30005385 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136644 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3150032MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 115350-24-8
Molekylær formel C12 H24 N2 O9 . 2(HCl)
Molekylvægt (g/mol) 413.25
Synonym D-Glucose, 2-amino-4-O-(2-amino-2-deoxy-β-D-glucopyranosyl)-2-deoxy-, hydrochloride (1:2) (ACI), 2-Amino-4-O-(2-amino-2-deoxy-β-D-glucopyranosyl)-2-deoxy-D-glucose hydrochloride (1:2) (ACI), D-Glucose, 2-amino-4-O-(2-amino-2-deoxy-β-D-glucopyranosyl)-2-deoxy-, dihydrochloride (9CI), Chitobiose dihydrochloride, 4-O-(2-Amino-2-deoxy-β-D-glucosyl)-D-glucosamine dihydrochloride
IUPAC navn (3R,4R,5S,6R)-3-amino-5-(((2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-2,4-diol dihydrochloride
SMIL Cl.Cl.N[C@H]1C(O)O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2N)[C@@H]1O
Tekniske data
Kemisk navn eller materiale Chitobiose Dihydrochloride
Mængde 2 mg
InChI formel InChI=1S/C12H24N2O9.2ClH/c13-5-9(19)10(4(2-16)21-11(5)20)23-12-6(14)8(18)7(17)3(1-15)22-12;;/h3-12,15-20H,1-2,13-14H2;2*1H/t3-,4-,5-,6-,7-,8-,9-,10-,11?,12+;;/m1../s1
Formel vægt 412.1