Få mere at vide
STEMCELL Technologies CHIR98014
Beskrivelse
- CHIR98014 is a reversible, cell-permeable activator of the WNT pathway, through inhibition of both isoforms of glycogen synthase kinase 3 (GSK3α and GSK3β) with IC50 values of 0.65 and 0.58 nM, respectively
- It shows at least 500-fold selectivity for GSK-3 versus 20 other serine/threonine or tyrosine kinases (Ring et al.)
Tekniske data
Tekniske data
| CAS | 556813-39-9 |
| Mængde | 1 mg |
| Molekylær formel | C20H17Cl2N9O2 |
| Renhed | ≥98% |