missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Description
CHIP/STUB1 Polyclonal specifically detects CHIP/STUB1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation.
Specifications
Specifications
| Applikationer | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Konjugeret | Alexa Fluor 750 |
| Fortynding | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulering | 50mM Sodium Borate |
| Værtsarter | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 253 and 303 of human STIP1 homology and U-box containing protein 1 (carboxy terminus of Hsp70-interacting protein) using the numbering given in entry NP_005852.2 (GeneID 10273). |
| Oprensningsmetode | Immunogen affinity purified |
| Mængde | 0.1 mL |
| Forskningsdisciplin | Neuroscience |
| Produkttype | Antibody |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?