missing translation for 'onlineSavingsMsg'
Få mere at vide

Thermo Scientific™ Bufret peptonvand (dehydreret)

Artikelnummer. 15862327
Klik for at se tilgængelige muligheder
Mængde:
500 g
2.5 kg
5 kg
25 kg
Pakningsstørrelse:
2.5kg
500g
5kg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 15862327

Brand: Thermo Scientific™ CM0509K

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Forberig fødevareprøver før isolering af Salmonella -arter, mens det tillader genoplivning af sub-dødeligt skadede celler med dette medium.

Forberig fødevareprøver før isolering af Salmonella -arter ved hjælp af Thermo Scientific™ Oxoid™ Bufret peptonvand (dehydreret). Bufret Peptone Water er et ikke-selektivt præberigelsesmedium. Det giver også betingelser for genoplivning af celler, der er blevet såret ved processer til konservering af fødevarer.

Low numbers and/or sub-lethally injured Salmonella spp. cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into a selective culture, improving the chances of recovering Salmonella species from the sample.

  • Supports high growth and recovery rate: rich in nutrients, the medium produces high resuscitation rates and supports intense growth.
  • Helps recovery of sub-lethally damaged salmonellae: the phosphate buffer system of the medium prevents bacterial damage due to changes in pH of the medium and helps the recovery of sub-lethally damaged salmonellae that may be sensitive to low pH.

It was noted by Edel and Kampelmacher1 that sub-lethal injury to salmonellae may occur in many food processes. In a survey involving isolation of salmonellae from meat that had been artificially contaminated with sub-lethally injured organisms, pre-enrichment in buffered peptone water at 37°C for 18 hours before selection in Brilliant Green Tetrathionate Bile Broth showed superior results compared with a direct selection method.

Sadovski2 reported that, in experiments involving isolation of salmonellae from frozen vegetables, the rapid drop in pH when using lactose broth3 as a pre-enrichment medium was detrimental to the recovery of salmonellae. This was due to the enhanced sensitivity to low pH of freeze-injured salmonellae which may contaminate frozen vegetables. Pre-enrichment with Buffered Peptone Water maintained a high pH over a period of 24 hours of incubation. The vegetable tissue has a low buffering capacity and the medium overcame this problem.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Tekniske data

Form Powder
Produkttype Pre-enrichment Medium
Mængde 25 kg

Kun til laboratoriebrug

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.