missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Chicken anti-Human IgG (H+L) Secondary Antibody, DyLight 350, Novus Biologicals™
Shop alle Bio Techne produkterDescription
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applikationer | Western Blot, ELISA, Immunohistochemistry, Dot Blot |
| Klassifikation | Polyclonal |
| Konjugeret | DyLight 350 |
| Formulering | 50mM Sodium Borate |
| Værtsarter | Chicken |
| Immunogen | Human IgG whole molecule |
| Oprensningsmetode | Multi-step |
| Mængde | 0.5 mL |
| Regulatorisk status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?