missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Chicken anti-Goat IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Shop alle Bio Techne produkterBeskrivelse
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay
Tekniske data
Tekniske data
| Antigen | IgG (H+L) |
| Applikationer | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Fortynding | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoassay, Immunomicroscopy 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA) |
| Formulering | 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 |
| Værtsarter | Chicken |
| Immunogen | This Chicken anti-Goat IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified goat IgG (H&L). |
| Oprensningsmetode | Affinity purified |
| Mængde | 0.5 mg |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?