missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Canine Hearthworm, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against Dirofilaria immitus.
Brand: Abnova PAB29148.100ug
Denne vare kan ikke returneres.
Se returpolitik
Tekniske data
| Canine Hearthworm | |
| Polyclonal | |
| Chicken polyclonal antibody raised against Dirofilaria immitus. | |
| In PBS, pH 7.2 (0.09% sodium azide). | |
| Extracts from both male and female heartworms (Dirofilaria immitus) with Freund's adjuvant. | |
| 100 μg | |
| Primary | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgY |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA(1:1000-1:10000) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| Chicken | |
| Affinity purification | |
| RUO | |
| Clostridium species | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion