missing translation for 'onlineSavingsMsg'
Få mere at vide

Chicago Sky Blue 6B, Thermo Scientific™

Artikelnummer. 11442367
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
100 g
500 g
Pakningsstørrelse:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11442367 500 g 500g
11422367 25 g 25g
11432367 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11442367 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A14242.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

C.I. 24410

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Chicago Sky Blue 6B is a counterstain for background autofluorescence in fluorescence and immunofluorescence histochemistry. It helps in the examination of blood plasma permeation.

Solubility
Soluble in water (1 mg/ml).

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2610-05-1
Molekylær formel C34H24N6Na4O16S4
Molekylvægt (g/mol) 992.79
MDL nummer MFCD00004020
InChI nøgle UPKAWFACSJWKND-ZXFFUEEESA-J
Synonym C.I. 24410; Direct Blue 1
PubChem CID 44135407
IUPAC navn tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methoxyphenyl]-2-methoxyphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate
SMIL COC1=C(C=CC(=C1)C2=CC(=C(C=C2)NN=C3C=CC4=C(C3=O)C(=C(C=C4S(=O)(=O)[O-])S(=O)(=O)[O-])N)OC)NN=C5C=CC6=C(C5=O)C(=C(C=C6S(=O)(=O)[O-])S(=O)(=O)[O-])N.[Na+].[Na+].[Na+].[Na+]

Tekniske data

Kemisk navn eller materiale Chicago Sky Blue 6B
Opløselighedsinformation Soluble in water (1mg/ml).
Formel vægt 992.79
Mængde 500 g
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.