Få mere at vide
Emma Safety Footwear Chicago Safety Shoes
Størrelse:
39
40
41
42
43
44
45
46
47
Pakningsstørrelse:
1 par
Beskrivelse
- S1P-Safety shoe with sole plate for dry work
- Knitted Textile construction
- Polyurethane coated
- Hydro-Tec™ Antibacterial
- Metal-free
- Chrome-free lining
- ESD Certified
Tekniske data
Tekniske data
| Certificeringer/compliance | EN ISO 20345:2011 |
| Lukketype | Laces |
| Farve | Gray, Black |
| Design | Chicago |
| Liner materiale | Hydro-Tec™ Antibacterial |
| Materiale | Textile |
| Størrelse | 40 |
| Tå materiale | Composite |