missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ CHIA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563681
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Immunogen sequence: DTGSNAYLNV DYVMNYWKDN GAPAEKLIVG FPTYGHNFIL SNPSNTGIGA PTSGAGPAGP YAKESGIWAY YE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
CHIA degrades chitin and chitotriose. May participate in the defense against nematodes and other pathogens. There are 3 named isoforms produced by alternative splicing. CHIA is induced via a T helper-2 (Th2)-specific, interleukin-13-mediated pathway in epithelial cells and macrophages. CHIA may be an important mediator of IL13-induced responses in Th2-dominated disorders such as asthma. CHIA hydrolysis N-acetyl-beta-D-glucosaminide 1,4-beta-linkages in chitin and chitodextrins.
Tekniske data
| CHIA | |
| Polyclonal | |
| Unconjugated | |
| CHIA | |
| 2200003E03Rik; acidic chitinase; acidic mammalian chitinase; AMCASE; Chia; Chia1; CHIT2; chitinase, acidic; chitinase, acidic 1; eosinophil chemotactic cytokine; lung-specific protein TSA1902; small acid mammalian chitinase; TSA1902; YNL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27159 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BZP6 | |
| CHIA | |
| Recombinant protein corresponding to Human CHIA. Recombinant protein control fragment (Product #RP-94208). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion