Få mere at vide
Chenopodium Oil, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Chenopodium Oil |
| CAS | 8006-99-3 |
| Mængde | 100 g |
| Synonym | Oil Of American Wormseed, Oil Of Chenopodium |