Få mere at vide
Chenodeoxycholic Acid Methyl Ester, TRC
Artikelnummer. 30358255
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30358255 250 mg -
30354042 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30358255 Leverandør Toronto Research Chemicals Leverandørnr. TRCC291905250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3057-04-3
Molekylær formel C25H42O4
Molekylvægt (g/mol) 406.6
Synonym (3α,5β,7α)-3,7-Dihydroxy-cholan-24-oic Acid Methyl Ester, Chenodeoxycholic Acid Methyl Ester, Methyl 3α,7α-Dihydroxy-5β-cholan-24-oate, Methyl 3α,7α-Dihydroxy-5β-cholanoate, Methyl Chenodeoxycholate
IUPAC navn methyl (4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMIL C[C@H](CCC(=O)OC)[C@H]1CC[C@H]2[C@H]3[C@H](CC[C@]12C)[C@@]4(C)CC[C@H](C[C@H]4C[C@H]3O)O
Tekniske data
Kemisk navn eller materiale Chenodeoxycholic Acid Methyl Ester
Mængde 250 mg
InChI formel InChI=1S/C25H42O4/c1-15(5-8-22(28)29-4)18-6-7-19-23-20(10-12-25(18,19)3)24(2)11-9-17(26)13-16(24)14-21(23)27/h15-21,23,26-27H,5-14H2,1-4H3/t15-,16+,17-,18-,19+,20+,21-,23+,24+,25-/m1/s1
Formel vægt 406.3083