Få mere at vide
Chenodeoxycholic Acid-d4, TRC
Artikelnummer. 30359345
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30359345 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30359345 Leverandør Toronto Research Chemicals Leverandørnr. TRCC29190250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chenodeoxycholic Acid-d4
CAS 99102-69-9
Renhedsgrad noter HPLC
Molekylær formel C24 D4 H36 O4
Mængde 50 mg
Synonym (3α,5β,7α)-3,7-Dihydroxycholan-24-oic Acid-d4, (+)-Chenodeoxycholic Acid-d4, 17β-(1-Methyl-3-carboxypropyl)etiocholane-3α,7α-diol-d4, Chenodeoxycholic Acid-d4, CDC-d4, Chendol-d4, Chenodiol-d4, Chenocol-d4, Fluibil-d4
InChI formel InChI=1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24-/m1/s1/i8D2,12D2
SMIL [2H]C1([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3[C@H](O)C[C@@H]2C([2H])([2H])[C@@H]1O)[C@H](C)CCC(=O)O
IUPAC navn (4R)-4-[(3R,5R,7R,8R,9S,10S,13R,14S,17R)-2,2,4,4-tetradeuterio-3,7-dihydroxy-10,13-dimethyl-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvægt (g/mol) 396.6
Formel vægt 396.3178
Procent renhed >95