Få mere at vide
Chenodeoxycholic Acid 24-Acyl-Beta-D-glucuronide, TRC
Artikelnummer. 30006527
Skift visning
Mængde:
2.5 mg
5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30006527 5 mg -
30003402 2.5 mg 2.5mg
30003751 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006527 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2919105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 208038-27-1
Molekylær formel C30H48O10
Molekylvægt (g/mol) 568.7
Synonym 1-[(3α,5β,7α)-3,7-Dihydroxycholan-24-oate] β-D-Glucopyranuronic Acid;
IUPAC navn (2S,3S,4S,5R,6S)-6-[(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMIL O[C@@H]1C[C@]2([H])C[C@H](O)CC[C@]2(C)[C@]3([H])[C@]1([H])[C@@]4([H])[C@]([C@@]([H])([C@H](C)CCC(O[C@H]5[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O5)=O)CC4)(C)CC3
Tekniske data
Kemisk navn eller materiale Chenodeoxycholic Acid 24-Acyl-beta-D-glucuronide
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C30H48O10/c1-14(4-7-21(33)39-28-25(36)23(34)24(35)26(40-28)27(37)38)17-5-6-18-22-19(9-11-30(17,18)3)29(2)10-8-16(31)12-15(29)13-20(22)32/h14-20,22-26,28,31-32,34-36H,4-13H2,1-3H3,(H,37,38)/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24+,25-,26+,28-,29+,30-/m1/s1
Procent renhed >95%