Få mere at vide
Chenodeoxycholic-2,2,4,4-d4 Acid, CDN
Artikelnummer. 30310848
Skift visning
Mængde:
0.05 g
Artikelnummer. Mængde
30310848 0.05 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310848 Leverandør CDN Leverandørnr. CDND27720.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chenodeoxycholic Acid-d4
CAS 99102-69-9
Molekylær formel C24 D4 H36 O4
Mængde 0.05 g
Synonym (3α,5β,7α)-3,7-Dihydroxycholan-24-oic Acid-d4, (+)-Chenodeoxycholic Acid-d4, 17β-(1-Methyl-3-carboxypropyl)etiocholane-3α,7α-diol-d4, Chenodeoxycholic Acid-d4, CDC-d4, Chendol-d4, Chenodiol-d4, Chenocol-d4, Fluibil-d4;
InChI formel InChI=1 S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26 H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24-/m1/s1/i8D2,12D2
SMIL [2 H]C1([2 H])C[C@]2(C)[C@H]3 CC[C@]4(C)[C@H](CC[C@H]4 [C@@H]3 [C@H](O)C[C@@H]2 C([2 H])([2 H])[C@@H]1 O)[C@H](C)CCC(=O)O
IUPAC navn (4 R)-4-[(3 R,5 R,7 R,8 R,9 S,10 S,13 R,14 S,17 R)-2,2,4,4-tetradeuterio-3,7-dihydroxy-10,13-dimethyl-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvægt (g/mol) 396.6
Formel vægt 396.3178 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity