Learn More
Cole-Parmer™ Stuart™ Chemically Resistant ECTFE Bottle Top Adaptor
Chemically resistant ECTFE bottle top adaptor compatible with the Stuart™ range of bottle top dispensers
Brand:  Cole-Parmer™ 10116-93
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12467762
  • 432.00 DKK / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • For use with a variety of Stuart™ bottle top dispensers
  • Chemically resistant ECTFE
  • Available in 38mm or 40mm thread size
Specifications
Specifications
Adapter
ECTFE
Pressmatic Bottle Top Dispenser
38 mm