missing translation for 'onlineSavingsMsg'
Få mere at vide

Chelidonic acid, MedChemExpress

Artikelnummer. 16449406
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16449406 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16449406 Leverandør Medchem Express Leverandørnr. HYW041489

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Chelidonic acid is a component of Chelidonium majus L., used as an antimicrobial. Chelidonic acid also shows anti-inflammatory activity. Chelidonic acid has potential to inhibit IL-6 production by blocking NF-κB and caspase-1. Chelidonic acid is a glutamate decarboxylase inhibitor, with a Ki of 1.2 μM.

Chelidonic acid is a component of Chelidonium majus L., used as an antimicrobial. Chelidonic acid also shows anti-inflammatory activity. Chelidonic acid has potential to inhibit IL-6 production by blocking NF-κB and caspase-1. Chelidonic acid is a glutamate decarboxylase inhibitor, with a Ki of 1.2 μM.

Tekniske data

Kemisk navn eller materiale Chelidonic acid
Fysisk form Solid
CAS 99-32-1
Farve Off-White to Gray
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C7H4O6
Til brug med (applikation) COVID-19-immunoregulation
Opløselighedsinformation DMSO : 25 mg/mL (135.80 mM; Need ultrasonic) ∣H2O : 2.5 mg/mL (13.58 mM; Need ultrasonic)
SMIL O=C(C1=CC(C=C(C(O)=O)O1)=O)O
Molekylvægt (g/mol) 184.1
Formel vægt 184.1
Procent renhed 95.41%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.