Få mere at vide
Tocris Bioscience™ CHDI 00390576
Beskrivelse
CHDI 00390576 is a potent and selective class IIa histone deacetylase (HDAC) inhibitor (IC50 values are 31, 50, 54 and 60 nM for HDAC7, HDAC9, HDAC4 and HDAC5, respectively). Displays >100-fold selectivity for class IIa HDACs over other HDACs. Brain penetrant.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-(2-Fluorophenyl)-N-hydroxy-2-(4-(5-(trifluoromethyl)pyrimidin-2-yl)phenyl)acetamide |
| CAS | 1629729-98-1 |
| Mængde | 5 mg |
| Molekylær formel | C19H13F4N3O2 |
| Renhed | 98% |