missing translation for 'onlineSavingsMsg'
Learn More

CGS 12066B dimaleate, 98+%, Thermo Scientific Chemicals

Catalog Number 15495969
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
109028-10-6
C25H25F3N4O8
566.49
MFCD00055048
HTEVMLYDEWVIQE-LVEZLNDCSA-N
cgs 12066b dimaleate, 1-methyl-4-7-trifluoromethyl pyrrolo 1,2-a quinoxalin-4-yl piperazine; bis butenedioic acid
71299720
(E)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline
CN1CCN(CC1)C2=NC3=C(C=CC(=C3)C(F)(F)F)N4C2=CC=C4.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O
This item is not returnable. View return policy
109028-10-6
C25H25F3N4O8
566.49
MFCD00055048
HTEVMLYDEWVIQE-LVEZLNDCSA-N
cgs 12066b dimaleate, 1-methyl-4-7-trifluoromethyl pyrrolo 1,2-a quinoxalin-4-yl piperazine; bis butenedioic acid
71299720
(E)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline
CN1CCN(CC1)C2=NC3=C(C=CC(=C3)C(F)(F)F)N4C2=CC=C4.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O

5-HT-1B full agonist

5-HT-1B full agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 126°C to 127°C
Color White
Quantity 50 mg
Formula Weight 566.49
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material CGS 12066B dimaleate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'