missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies CGP77675
Beskrivelse
- CGP77675 is a potent inhibitor of Src family kinases
- It has been reported to inhibit phosphorylation of peptide substrates and autophosphorylation of purified Src (IC50 = 5 - 20 nM and 40 nM, respectively), Src, and Src kinase family members such as LCK (IC50 = 0.02 mM and 0.29 mM, respectively)
- It has also been shown to inhibit epidermal growth factor receptor (EGFR), Cdc2, v-Abl, and focal adhesion kinase (FAK) (IC50 = 0.15 mM, > 10 mM, 0.31 mM, and 0.2 mM, respectively; Missbach et al.)
- In combination with the GSK3 inhibitor CHIR99021 (Catalog #72052), CGP77675 is used to maintain mouse embryonic stem (ES) cells (Shimizu et al.)
- Preserves the epigenetic and genomic integrity as well as the developmental potential of ES cells (Choi et al.)
Tekniske data
Tekniske data
| CAS | 234772-64-6 |
| Mængde | 5 mg |
| Molekylær formel | C26H29N5O2 |
| Renhed | ≥98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?