Få mere at vide
Tocris Bioscience™ CGI 1746
Beskrivelse
CGI 1746 is a potent, reversible inhibitor of Bruton's tyrosine kinase (Btk) (IC50 = 1.9 nM). CGI 1746 decreases cell survival in prostate cancer cell lines. In a cell model, CGI 1746 blocks the production of FcγR-induced cytokines IL-1β, TNFα and IL-6 (IC50 values are 36 nM, 47 nM and 353 nM, respectively).
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[3-[4,5-Dihydro-4-methyl-6-[[4-(4-morpholinylcarbonyl)phenyl]amino]-5-oxo-2-pyrazinyl]-2-methylphenyl]-4-(1,1-dimethylethyl)benzamide |
| CAS | 910232-84-7 |
| Mængde | 10 mg |
| Mål | Brutons Tyrosine Kinase (BTK) Inhibitors |
| Molekylær formel | C34H37N5O4 |
| Renhed | 0.97 |