missing translation for 'onlineSavingsMsg'
Få mere at vide

Cevimeline hydrochloride hemihydrate, MedChemExpress

Artikelnummer. 16468218
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16468218 10 mg
16444117 50 mg
16440658 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16468218 Leverandør Medchem Express Leverandørnr. HY76772

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Cevimeline hydrochloride hemihydrate (SNI-2011) is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. Cevimeline hydrochloride hemihydrate stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. Cevimeline hydrochloride hemihydrate can cross the blood-brain barrier (BBB).

Cevimeline hydrochloride hemihydrate (SNI-2011) is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. Cevimeline hydrochloride hemihydrate stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. Cevimeline hydrochloride hemihydrate can cross the blood-brain barrier (BBB).

Tekniske data

Kemisk navn eller materiale Cevimeline hydrochloride hemihydrate
Fysisk form Solid
CAS 153504-70-2
Farve White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C10H17NOS.HCl.1/2H2O
Til brug med (applikation) Neuroscience-Neuromodulation
Synonym SNI-2011AF102B hydrochloride hemihydrate
Opløselighedsinformation H2O : ≥ 50 mg/mL (204.27 mM)
SMIL C[C@H]1SC[C@@]2(CN3CCC2CC3)O1.[H]Cl.[Relative Stereochemistry].[0.5 H2O]
Molekylvægt (g/mol) 244.78
Formel vægt 244.78
Procent renhed 95.0%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.