Få mere at vide
Cetyltrimethylammonium Hydrogensulfate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Cetyltrimethylammonium Hydrogensulfate |
| CAS | 68214-07-3 |
| Molekylær formel | C19 H42 N . H O4 S |
| Mængde | 5 g |
| Synonym | 1-Hexadecanaminium, N,N,N-trimethyl-, sulfate (1:1) (9CI, ACI), Cetyltrimethylammonium hydrogen sulfate, CTAHSO4, Hexadecyltrimethylammonium bisulfate, Hexadecyltrimethylammonium hydrogen sulfate, Hexadecyltrimethylammonium hydrogensulfate, Cetrimonium hydrogen sulfate |
| InChI formel | InChI=1S/C19H42N.H2O4S/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;1-5(2,3)4/h5-19H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1 |
| SMIL | CCCCCCCCCCCCCCCC[N+](C)(C)C.OS(=O)(=O)[O-] |
| IUPAC navn | hexadecyl(trimethyl)ammonium;hydrogen sulfate |
| Molekylvægt (g/mol) | 381.61 |
| Formel vægt | 381.2913 |