Få mere at vide
Cetylpyridinium chloride monohydrate, MedChemExpress
Artikelnummer. 16498716
Encompass_Preferred
Skift visning
Mængde:
100 mg
Pakningsstørrelse:
100mg
Artikelnummer. Mængde unitSize
16498716 100 mg 100mg
1 options
Artikelnummer. 16498716 Leverandør Medchem Express Leverandørnr. HYB1289
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
MedChemExpress Cetylpyridinium chloride monohydrate is a cationic quaternary ammonium compound, used in some types of mouthwashes, toothpastes, throat and nasal sprays, is an antiseptic that kills bacteria and other microorganisms, effective in preventing dental plaque and reducing gingivitis.
Cetylpyridinium chloride monohydrate is a cationic quaternary ammonium compound, used in some types of mouthwashes, toothpastes, throat and nasal sprays, is an antiseptic that kills bacteria and other microorganisms, effective in preventing dental plaque and reducing gingivitis.
Tekniske data
Kemisk navn eller materiale Cetylpyridinium chloride monohydrate
Fysisk form Powder
CAS 6004-24-6
Farve Off-White
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C21H40ClNO
Til brug med (applikation) COVID-19-immunoregulation
Synonym Hexadecylpyridinium chloride monohydrate
Opløselighedsinformation H2O : ≥ 100 mg/mL (279.33 mM) ∣DMSO : 20 mg/mL (55.87 mM; Need ultrasonic)
SMIL CCCCCCCCCCCCCCCC[N+]1=CC=CC=C1.O.[Cl-]
Molekylvægt (g/mol) 358.0
Formel vægt 358.0
Procent renhed 99.79%