Få mere at vide
Cetyl Stearate, TRC
Artikelnummer. 30358248
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30358248 5 g
30356587 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30358248 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2816005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1190-63-2
Molekylær formel C34 H68 O2
Molekylvægt (g/mol) 508.9
Synonym Octadecanoic Acid Hexadecyl Ester, Stearic Acid Hexadecyl Ester, 1-Hexadecanol Stearate, Hexadecyl Octadecanoate, Hexadecyl Stearate, Palmityl Stearate, Wickenol 121, n-Hexadecyl Stearate
IUPAC navn hexadecyl octadecanoate
SMIL CCCCCCCCCCCCCCCCCC(=O)OCCCCCCCCCCCCCCCC
Tekniske data
Kemisk navn eller materiale Cetyl Stearate
Mængde 5 g
InChI formel InChI=1S/C34H68O2/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34(35)36-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2/h3-33H2,1-2H3
Formel vægt 508.5219