Få mere at vide
Cetoxime Hydrochloride, LoGiCal
Artikelnummer. 30460426
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460426 10 mg
1 options
Artikelnummer. 30460426 Leverandør LoGiCal Leverandørnr. LGCFOR1660.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cetoxime Hydrochloride
CAS 22204-29-1
Analyt- eller komponentnavne Cetoxime Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C15 H17 N3 O . Cl H
Certificeringer/compliance ISO 9001
Synonym Ethanimidamide, N-hydroxy-2-[phenyl(phenylmethyl)amino]-, hydrochloride (1:1), Acetamidoxime, 2-(N-benzylanilino)-, hydrochloride, Acetamidoxime, 2-(N-benzylanilino)-, monohydrochloride, Ethanimidamide, N-hydroxy-2-[phenyl(phenylmethyl)amino]-, monohydrochloride, Cetoxime hydrochloride, Febramine
InChI formel InChI=1S/C15H17N3O.ClH/c16-15(17-19)12-18(14-9-5-2-6-10-14)11-13-7-3-1-4-8-13;/h1-10,19H,11-12H2,(H2,16,17);1H
Løsningstype Neat
SMIL Cl.N\C(=N/O)\CN(Cc1ccccc1)c2ccccc2
IUPAC navn 2-(N-benzylanilino)-N'-hydroxyethanimidamide;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 291.78
Mængde 10 mg
Formel vægt 291.1138 g/mol