Få mere at vide
(±)-Cetirizine-d4 (ethoxy-d4), CDN
Artikelnummer. 30318804
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318804 0.005 g
30312377 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318804 Leverandør CDN Leverandørnr. CDND61000.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-Cetirizine-d4 (ethoxy-d4)
CAS 1219803-84-5
Molekylær formel C21 D4 H21 Cl N2 O3
Mængde 0.005 g
Synonym 2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]-1,1,2,2-tetradeuterioethoxy]acetic acid, 2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy-1,1,2,2-d4]acetic acid, Cetirizine-d4, Cetirizine-D4 (ethoxy-1,1,2,2-D4), (±)-Cetirizine-d4 (ethoxy-d4)
InChI formel InChI=1 S/C21H25ClN2O3/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26/h1-9,21 H,10-16H2,(H,25,26)/i14D2,15D2
SMIL [2 H]C([2 H])(OCC(=O)O)C([2 H])([2 H])N1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC navn 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]-1,1,2,2-tetradeuterioethoxy]acetic acid
Molekylvægt (g/mol) 392.9124
Formel vægt 392.1805 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity