Få mere at vide
Cetirizine 2-Chloro Impurity Dihydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (RS)-2-[2-[4-[(2-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride |
| CAS | 2702511-37-1 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C21 H25 Cl N2 O3 . 2 Cl H |
| Mængde | 10 mg |
| Synonym | (RS)-2-[2-[4-[(2-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride, Cetirizine Dihydrochloride Imp. C (EP) as Dihydrochloride |
| InChI formel | InChI=1S/C21H25ClN2O3.2ClH/c22-19-9-5-4-8-18(19)21(17-6-2-1-3-7-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26;;/h1-9,21H,10-16H2,(H,25,26);2*1H |
| SMIL | Cl.Cl.OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccccc3Cl |
| IUPAC navn | 2-[2-[4-[(2-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride |
| Molekylvægt (g/mol) | 461.81 |
| Vis mere |