Learn More
Cesium Nitrate, 99%+, MilliporeSigma™
Product Code. 15854310
Change view
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15854310 1 kg Plastic bottle 1kg
15844310 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15854310 Supplier Supelco Supplier No. 1028561000
 Restricted Item
Forventet forsendelse: 26-11-2026
High-purity cesium nitrate designed for analytical applications
  • High purity chemical for analysis in research and laboratory
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.909
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
Synonym cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]
Specifications
Melting Point 401°C
Color Off-white
pH 5.0 to 7.5
Physical Form Solid
Quantity 1 kg
Assay Percent Range 99%+
MDL Number MFCD00010963
UN Number UN 1451
Solubility Information 239.5 g/L in Water
InChI Formula 1S/Cs.NO3/c;2-1(3)4/q+1;-1
Formula Weight 194.91
Grade Analytical
Odor Odorless
Packaging Plastic bottle
Product Line EMSURE™
Appearance Off-white solid
Density 3.64g/cm³
Chemical Name or Material Cesium nitrate