missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride, 99.0%, (Reagent Grade), MP Biomedicals
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
Specifications
Quantity | 25 g |
Assay Percent Range | 99.0% |
IUPAC Name | caesium(1+) chloride |
Formula Weight | 168.4 |
Grade | Reagent |
Chemical Name or Material | Cesium Chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'