Learn More
Certified Reference Flash Point Standard - PMCC Lubricant, UKAS ISO 17034 Certified 250mL (nominal value: 190.5 °C), Paragon Scientific™
Flash Point Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Flash point according to ASTM D93.
Brand: Paragon Scientific CRMU-PMLU
This item is not returnable.
View return policy
Description
- Certified Reference Material for annual verification checks in accordance with ASTM D93.
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Certified Reference Flash Point Standard - PMCC Lubricant, UKAS ISO 17034 Certified 250mL (nominal value: 190.5 °C), Paragon Scientific™ > Quantity: 250mL
Nominal values quoted. Certified Values will vary from batch to batch.
Spot an opportunity for improvement?Share a Content Correction