missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(Iii) Nitrate Hexahydrate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | Ce(NO3)3 · 6H2O |
UN Number | 1477 |
Formula Weight | 434.22g/mol |
Percent Purity | ≥99% |
Grade | puriss. p.a. |
Packaging | Plastic Bottle |
Chemical Name or Material | Cerium(Iii) Nitrate Hexahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'