Få mere at vide
Tocris Bioscience™ Cerestat
Potent and noncompetitive NMDA antagonist
Brand: Tocris Bioscience™ 4492/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Cerestat is a potent and noncompetitive NMDA receptor antagonist (IC50 = 36 nM). Exhibits >70-fold selectivity for NMDA receptor over σ receptor. Neuroprotective in a rat model of focal ischemia.Tekniske data
| Cerestat | |
| N-(3-Ethylphenyl)-N-methyl-N'-1-naphthalenylguanidine hydrochloride | |
| Soluble to 50mM in water and to 100mM in DMSO | |
| Bioactive Small Molecule |
| 137160-11-3 | |
| 10 mg | |
| Solid |
Produkttitel
Ser du en mulighed for forbedring?
Research Use Only. Not For Human or Veterinary Use