missing translation for 'onlineSavingsMsg'
Få mere at vide

Cepharanthine, TRC

Artikelnummer. 30362737
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362737 100 mg
30366962 1 g
30367882 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30362737 Leverandør Toronto Research Chemicals Leverandørnr. TRCC261513100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 481-49-2
Molekylær formel C37 H38 N2 O6
Molekylvægt (g/mol) 606.71
Synonym 1H-4,6:16,19-Dietheno-21,25-metheno-12H-[1,3]dioxolo[4,5-g]pyrido[2',3':17,18][1,10]dioxacycloeicosino[2,3,4-ij]isoquinoline, 2,3,13,14,14a,15,26,26a-octahydro-22,30-dimethoxy-1,14-dimethyl-, (14aS,26aR)-, Cepharanthine (6CI,7CI,8CI), Oxyacanthan, 6',12'-dimethoxy-2,2'-dimethyl-6,7-[methylenebis(oxy)]-, (+)-Cepharanthine, 1H-4,6:16,19-Dietheno-21,25-metheno-12H-[1,3]dioxolo[4,5-g]pyrido[2',3':17,18][1,10]dioxacycloeicosino[2,3,4-ij]isoquinoline, 2,3,13,14,14a,15,26,26a-octahydro-22,30-dimethoxy-1,14-dimethyl-, [14aS-(14aR*,26aS*)]-, Cepharanthin, Cepharantin, NSC 623442, O-Methylcepharanoline
IUPAC navn (14S,27R)-22,33-dimethoxy-13,28-dimethyl-2,5,7,20-tetraoxa-13,28-diazaoctacyclo[25.6.2.216,19.13,10.121,25.04,8.031,35.014,39]nonatriaconta-1(33),3(39),4(8),9,16(38),17,19(37),21,23,25(36),31,34-dodecaene
SMIL COc1ccc2C[C@H]3N(C)CCc4cc(OC)c(Oc5c6OCOc6cc7CCN(C)[C@@H](Cc8ccc(Oc1c2)cc8)c57)cc34

Specifications

Kemisk navn eller materiale Cepharanthine
Mængde 100 mg
InChI formel InChI=1S/C37H38N2O6/c1-38-13-11-24-18-31(41-4)33-20-27(24)28(38)16-23-7-10-30(40-3)32(17-23)44-26-8-5-22(6-9-26)15-29-35-25(12-14-39(29)2)19-34-36(37(35)45-33)43-21-42-34/h5-10,17-20,28-29H,11-16,21H2,1-4H3/t28-,29+/m1/s1
Formel vægt 606.273
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.