Få mere at vide
Cephalothin Sodium, TRC
Artikelnummer. 30133904
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133904 100 mg
30132884 1 g
17933825 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133904 Leverandør Toronto Research Chemicals Leverandørnr. TRCC261155100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 58-71-9
Molekylær formel C16 H15 N2 O6 S2 . Na
Molekylvægt (g/mol) 418.42
Synonym Cefalotin sodium, Sodium (6R,7R)-3-[(acetyloxy)methyl]-8-oxo-7-[(thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-8-oxo-7-[[2-(2-thienyl)acetyl]amino]-, sodium salt (1:1), (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-(hydroxymethyl)-8-oxo-7-[2-(2-thienyl)acetamido]-, acetate (ester), monosodium salt (8CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-8-oxo-7-[(2-thienylacetyl)amino]-, monosodium salt, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[(acetyloxy)methyl]-8-oxo-7-[(2-thienylacetyl)amino]-, monosodium salt, (6R-trans)-, 38253, Averon 1, Cefalothin sodium, Cefalothine sodium, Cemastin, Cephalothin sodium, Cephalothin sodium salt, Cephation, Ceporacin, Cepovenin, Coaxin, Keflin, Lilly 38253, Lospoven, Microtin, Seffin, Sodium (thienylacetamido)cephalosporanate, Sodium 3-acetoxymethyl-7β-(2-thienylacetamido)ceph-3-em-4-carboxylate, Sodium 7-(2-thienylacetamido)-3-acetoxymethyl-3-cephem-4-carboxylate, Sodium 7-[2-(2-thienyl)acetamido]cephalosporanate, Sodium cefalotin, Sodium cephalothin, Sodium cephalotin, Synclotin, Toricelocin
IUPAC navn sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMIL [Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)Cc3cccs3)C2=O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale Cefalotin Sodium
Mængde 100 mg
InChI formel InChI=1S/C16H16N2O6S2.Na/c1-8(19)24-6-9-7-26-15-12(14(21)18(15)13(9)16(22)23)17-11(20)5-10-3-2-4-25-10;/h2-4,12,15H,5-7H2,1H3,(H,17,20)(H,22,23);/q;+1/p-1/t12-,15-;/m1./s1
Formel vægt 418.0269