Få mere at vide
Cephaeline dihydrochloride, MedChemExpress
Beskrivelse
Cephaeline dihydrochloride is a selective CYP2D6 inhibtor with an IC50 of 121 μM.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Cephaeline dihydrochloride |
| CAS | 5853-29-2 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C28H40Cl2N2O4 |
| Mængde | 5 mg |
| Synonym | (-)-Cephaeline dihydrochloride NSC 32944 |
| Opløselighedsinformation | DMSO : 250 mg/mL (463.37 mM; Need ultrasonic) ∣H2O : 125 mg/mL (231.68 mM; Need ultrasonic) |
| SMIL | Cl[H].CC[C@@H](CN1CC2)[C@H](C[C@@]1([H])C3=C2C=C(OC)C(OC)=C3)C[C@]4([H])C5=CC(OC)=C(O)C=C5CCN4.Cl[H] |
| Molekylvægt (g/mol) | 539.53 |
| Vis mere |