Få mere at vide
CEP57 (Human) Matched Antibody Pair, Polyclonal , Abnova™
Artikelnummer. 16103044
Skift visning
Mængde:
1 Set
Pakningsstørrelse:
1 sæt
Artikelnummer. Mængde unitSize
16103044 1 Set 1 sæt
1 options
Artikelnummer. 16103044 Leverandør Abnova Leverandørnr. H00009702AP52.1Set
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
Capture antibody: mouse monoclonal anti-CEP57 (100μg)
Detection antibody: rabbit purified polyclonal anti-CEP57 (50μg)
Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CEP57.
Translokin binds basic fibroblast growth factor (FGF2; MIM 134920) and mediates its nuclear translocation and mitogenic activity (Bossard et al., 2003 [PubMed 12717444]).[supplied by OMIM]
Tekniske data
Antigen CEP57
Applikationer ELISA
Klassifikation Monoclonal/Polyclonal
Konjugeret Unconjugated
Oversigt This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CEP57.
Sygdomstilstand Cardiovascular Diseases, Diabetes Mellitus Type 2, Edema
Gene CEP57
Gene Alias KIAA0092/PIG8/TSP57
Gensymboler CEP57
Mængde 1 Set
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 9702
Inkluderer Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CEP57 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-CEP57 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Målarter Human
Indhold og opbevaring Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Purified