Få mere at vide
Centrinone, TRC
Artikelnummer. 30348953
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30348953 1 mg -
30356103 2.5 mg 2.5mg
30352381 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30348953 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2566131MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1798871-30-3
Molekylær formel C26H25F2N7O6S2
Molekylvægt (g/mol) 633.65
Synonym 2-[[2-Fluoro-4-[[(2-fluoro-3-nitrophenyl)methyl]sulfonyl]phenyl]thio]-5-methoxy-N-(5-methyl-1H-pyrazol-3-yl)-6-(4-morpholinyl)-4-pyrimidinamine
IUPAC navn 2-[2-fluoro-4-[(2-fluoro-3-nitrophenyl)methylsulfonyl]phenyl]sulfanyl-5-methoxy-N-(5-methyl-1H-pyrazol-3-yl)-6-morpholin-4-ylpyrimidin-4-amine
SMIL FC1=C(C=CC=C1CS(C2=CC=C(C(F)=C2)SC3=NC(N4CCOCC4)=C(C(NC5=NNC(C)=C5)=N3)OC)(=O)=O)[N+]([O-])=O
Tekniske data
Kemisk navn eller materiale Centrinone
Mængde 1 mg
InChI formel InChI=1S/C26H25F2N7O6S2/c1-15-12-21(33-32-15)29-24-23(40-2)25(34-8-10-41-11-9-34)31-26(30-24)42-20-7-6-17(13-18(20)27)43(38,39)14-16-4-3-5-19(22(16)28)35(36)37/h3-7,12-13H,8-11,14H2,1-2H3,(H2,29,30,31,32,33)