missing translation for 'onlineSavingsMsg'
Få mere at vide

Applied Biosystems™ Centri-Sep™ 96-Well Plates

Artikelnummer. 16385302 Shop alle Applied Biosystems produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2 tallerkener
50 tallerkener
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16385302 2 tallerkener
16370824 50 tallerkener
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16385302 Leverandør Applied Biosystems™ Leverandørnr. 4367819

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

Indeholder

This kit contains two Centri-Sep™ 96 plates, two 96-well PCR collection plates, and two sheets of ThermalSeal film

Centri-Sep™ 96's hydrated matrix enables effective and reliable high-throughput removal of excess BigDye™ terminators from DNA sequencing reactions, giving you clean sequencing reads with uniform signal intensity starting with the first base.

  • Processed faster than ultrafiltration columns
  • Rapid and efficient separations
  • Buffer is not preselected
  • Convenient 20 to 100μL sample size
  • Dry gel format
  • Room Temperature Storage
  • Microcentrifuge format

Ordreinfo

96-well Plate

TRUSTED_SUSTAINABILITY

Tekniske data

Indhold og opbevaring This kit contains two Centri-Sep™ 96 plates, two 96-well PCR collection plates, and two sheets of ThermalSeal film.
Format 96-well Plate
Prøvetype Sequencing Products
Til brug med (applikation) Sequencing
Produktlinje Centri-Sep
Produkttype 96-Well Plate
Mængde 2 tallerkener
Forsendelsestilstand Wet Ice
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.