Indeholder
This kit contains 50 Centri-Sep™ 96 plates
- Processed faster than ultrafiltration columns
- Rapid and efficient separations
- Buffer is not preselected
- Convenient 20 to 100μL sample size
- Dry gel format
- Room Temperature Storage
- Microcentrifuge format
Tekniske data
Tekniske data
| Indhold og opbevaring | This kit contains 50 Centri-Sep™ 96 plates. |
| Format | 96-well Plate |
| Produktlinje | Centri-Sep |
| Produkttype | 96-Well Plate |
| Mængde | 50 tallerkener |
| Forsendelsestilstand | Wet Ice |