Få mere at vide
Tocris Bioscience™ CELT-426
Artikelnummer. 18005372
Mængde:
10 μg
Artikelnummer. 18005372
Brand: Tocris Bioscience™ 7955/10U
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Potent and partially selective hD2 dopamine receptor fluorescent antagonist
CELT-426 is a potent and partially selective fluorescent hD2 dopamine receptor antagonist (Ki values are 89.3 nM, 194.8 nM, and 263.6 nM for D2, D3, and D4 dopamine receptors respectively in radioligand binding assay). CELT-426 can be used as a fluorescent dopamine receptor probe for fluorescence binding assays, live cell imaging, flow cytometry, and fluorescence polarization assays. It is a suitable replacement for GPCR radiolabeling ligands. Excitation and emission maxima (λ) are 560 and 571 nm, respectively. Wavelengths are compatible with use as an acceptor dye in TR-FRET assays, together with the CoraFluor™ 1 (Cat. No. 7920) TR-FRET donor. Add 62 μL of assay buffer containing 1% of DMSO to obtain a 100 μM stock solution, and use 250 nM as working concentration.
TRUSTED_SUSTAINABILITY
Tekniske data
Mængde 10 μg
Mål Dopamine D2R/DRD2
Renhed 95%