Få mere at vide
R&D Systems™ CellXVivo Human Monocyte-derived DC Differentiation Kit
Artikelnummer. 18707707
Skift visning
Mængde:
1 sæt
Artikelnummer. Mængde
18707707 1 sæt
1 options
Artikelnummer. 18707707 Leverandør R&D Systems™ Leverandørnr. CDK004
 Begrænset vare
Forventet forsendelse: 26-11-2026
Indeholder
Serum-Free Dendritic Cell Base Media; Recombinant Human IL-4; Recombinant Human GM-CSF; Recombinant Human TNF-alpha; Reconstitution Buffer 2
For the differentiation of monocyte-derived dendritic cells
For the differentiation of CD14+ monocytes into dendritic cells.
TRUSTED_SUSTAINABILITY
Tekniske data
Produktlinje R&D Systems
Mængde 1 sæt
Indhold og opbevaring Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.