Learn More
Sartorius™ Cellulose / Polyethylene Coated Surface Protection Paper
Product Code. 11734986 Shop All Sartorius Products
Change view
Click to view available options
Length (Metric):
50 m
100 m
Width (Metric):
400 mm
460 mm
600 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
11734986 50 m 460 mm Each
10140851 100 m 600 mm Each
10706701 50 m 400 mm Each
11724986 100 m 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11734986 Supplier Sartorius™ Supplier No. FT1601460050
  • ONLINE EXCLUSIVE PRICE
    1645.00 DKK
    1224.50 DKK / Pakke med 1
    Spare 420.50 DKK
    25% Slukket
Forventet forsendelse: 23-12-2026
Specifications
Product Type Surface Protection Paper
Material Cellulose/Polyethylene
Absorbancy 125 mL/m2
Length (Metric) 50 m
Width (Metric) 460 mm
Color White