Learn More
Nexcelom™ Cellometer SD025 Counting Chambers
3 ÎĽL volume load
Brand:  Nexcelom™ CHT4-SD025-002
Encompass
Product Code. 15829354
  • POA / Pack of 75
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
  • Each all-plastic, disposable slide contains two sample counting chambers with precisely controlled height
  • After loading the sample, the slide is inserted into a Cellometer System
  • Cells are imaged directly from the center of the counting chamber
  • Image-based counting with disposable counting chambers offers several key advantages:
    • No potential clogging
    • Ideal for fragile samples, such as hepatocytes
    • No washing
    • No potential cross-contamination
Specifications
Specifications
Slide
75 slides for 150 counts
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Nexcelom™ Cellometer SD025 Counting Chambers