missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CellAdhere™ Laminin-521

Matrix for maintenance of human ES and iPS cells in combination with mTeSR™1, TeSR™-E8™, or TeSR™2

Encompass_Preferred

POA - 961.20 DKK

Specifications

Cell Type Pluripotent Stem Cells
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15834087
Encompass_Preferred
View Documents
STEMCELL Technologies
77004
10 x 100 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15844755
Encompass_Preferred
View Documents
STEMCELL Technologies
77003
100 ÎĽg
961.20 DKK
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Support the growth and differentiation of human embryonic stem (ES) and induced pluripotent stem (iPS) cells under feeder-free conditions with this defined and xeno-free cell culture matrix
  • For consistent cell populations and reproducible results in downstream applications, use CellAdhere™ Laminin-521 with TeSR™ maintenance media to provide a defined culture substrate for cell maintenance
  • Laminin 521 is expressed and secreted by human pluripotent stem cells (hPSCs) in the inner cell mass of the embryo and therefore creates a biologically relevant hPSC culture environment in vitro
  • Compared to other matrices, CellAdhere™ Laminin-521 increases single-cell attachment and survival and does not require the addition of apoptotic inhibitors during plating
  • Note that single-cell passaging of human ES and iPS cells can result in selective pressure and lead to genetic aberrations
  • If passaging as single cells, we recommend checking the karyotype frequently
Specifications

Specifications

Pluripotent Stem Cells
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
STEMCELL Technologies CellAdhere™ Laminin-521

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.