Få mere at vide
Celiprolol Hydrochloride, LoGiCal
Artikelnummer. 30464059
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464059 10 mg
1 options
Artikelnummer. 30464059 Leverandør LoGiCal Leverandørnr. LGCFOR0128.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Celiprolol Hydrochloride
CAS 57470-78-7
Analyt- eller komponentnavne Celiprolol Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C20 H33 N3 O4 . Cl H
Certificeringer/compliance ISO 9001
Synonym Celiprolol Hydrochloride, Celectol, Corliprol, NSC 324509, REV 5320A, ST 1396, Selecor, Urea, N'-[3-acetyl-4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-N,N-diethyl-, monohydrochloride (9CI)
InChI formel InChI=1S/C20H33N3O4.ClH/c1-7-23(8-2)19(26)22-15-9-10-18(17(11-15)14(3)24)27-13-16(25)12-21-20(4,5)6;/h9-11,16,21,25H,7-8,12-13H2,1-6H3,(H,22,26);1H
Løsningstype Neat
SMIL Cl.CCN(CC)C(=O)Nc1ccc(OCC(O)CNC(C)(C)C)c(c1)C(=O)C
IUPAC navn 3-[3-acetyl-4-[3-(tert-butylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 415.95
Mængde 10 mg
Formel vægt 415.2238 g/mol