Få mere at vide
Celiprolol Hydrochloride EP Impurity F, Mikromol™
Artikelnummer. 30049479
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049479 25 mg
30052602 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049479 Leverandør Mikromol Leverandørnr. MM0128.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Celiprolol Hydrochloride EP Impurity F
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(3-Acetyl-4-hydroxyphenyl)-1,1-diethylurea
CAS 79881-89-3
Analyt- eller komponentnavne 3-(3-Acetyl-4-hydroxyphenyl)-1,1-diethylurea
Forsendelsestilstand Room Temperature
Molekylær formel C13 H18 N2 O3
Certificeringer/compliance ISO 17025
Synonym Celiprolol Hydrochloride Imp. F (EP), Celiprolol Imp. F (EP), 3-(3-Acetyl-4-hydroxyphenyl)-1,1-diethylurea, Celiprolol Hydrochloride Impurity F, Celiprolol Impurity F
InChI formel InChI=1S/C13H18N2O3/c1-4-15(5-2)13(18)14-10-6-7-12(17)11(8-10)9(3)16/h6-8,17H,4-5H2,1-3H3,(H,14,18)
SMIL CCN(CC)C(=O)Nc1ccc(O)c(c1)C(=O)C
IUPAC navn 3-(3-acetyl-4-hydroxyphenyl)-1,1-diethylurea
Molekylvægt (g/mol) 250.29
Mængde 25 mg
Formel vægt 250.1317 g/mol