Få mere at vide
Celiprolol EP Impurity C, Mikromol™
Artikelnummer. 30050371
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050371 100 mg
30050307 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050371 Leverandør Mikromol Leverandørnr. MM0128.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Celiprolol EP Impurity C
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[3-Acetyl-4-[(2RS)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-3-(1,1-dimethylethyl)urea
CAS 57471-01-9
Analyt- eller komponentnavne 1-[3-Acetyl-4-[(2RS)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-3-(1,1-dimethylethyl)urea
Forsendelsestilstand Room Temperature
Molekylær formel C20 H33 N3 O4
Certificeringer/compliance ISO 17025
Synonym Celiprolol Hydrochloride Imp. C (EP), Celiprolol Imp. C (EP), 1-[3-Acetyl-4-[(2RS)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]-3-(1,1-dimethylethyl)urea, Celiprolol Hydrochloride Impurity C, Celiprolol Impurity C
InChI formel InChI=1S/C20H33N3O4/c1-13(24)16-10-14(22-18(26)23-20(5,6)7)8-9-17(16)27-12-15(25)11-21-19(2,3)4/h8-10,15,21,25H,11-12H2,1-7H3,(H2,22,23,26)
SMIL CC(=O)c1cc(NC(=O)NC(C)(C)C)ccc1OCC(O)CNC(C)(C)C
IUPAC navn 1-[3-acetyl-4-[3-(tert-butylamino)-2-hydroxypropoxy]phenyl]-3-tert-butylurea
Molekylvægt (g/mol) 379.49
Mængde 100 mg
Formel vægt 379.2471 g/mol