Få mere at vide
Celiprolol-d9 Hydrochloride, TRC
Artikelnummer. 30353865
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30353865 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353865 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2545021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Celiprolol-d9 Hydrochloride
CAS 1215535-20-8
Renhedsgrad noter HPLC
Molekylær formel C20H25D9ClN3O4
Mængde 1 mg
Synonym N'-[3-Acetyl-4-[3-[(1,1-dimethylethyl-d9)amino]-2-hydroxypropoxy]phenyl]-N,N-diethylurea Hydrochloride, Celectol-d9, Corliprol-d9, NSC 324509-d9, REV 5320A-d9, ST 1396-d9, Selecor-d9
InChI formel InChI=1S/C20H33N3O4.ClH/c1-7-23(8-2)19(26)22-15-9-10-18(17(11-15)14(3)24)27-13-16(25)12-21-20(4,5)6;/h9-11,16,21,25H,7-8,12-13H2,1-6H3,(H,22,26);1H/i4D3,5D3,6D3;
SMIL CCN(CC)C(NC1=CC(C(C)=O)=C(OCC(O)CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1)=O.Cl
IUPAC navn 3-[3-acetyl-4-[3-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]-2-hydroxypropoxy]phenyl]-1,1-diethylurea;hydrochloride
Molekylvægt (g/mol) 425.01
Procent renhed >95